Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Methyl-2-pyridone ≥98%

4-Methyl-2-pyridone ≥98%

4-Methyl-2-pyridone ≥98%
Certificates
4-Methyl-2-pyridone ≥98%
4-Methyl-2-pyridone ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 13466-41-6 Molecular Formula: C6H7NO Formula Weight: 109.13 Odour: Odourless Melting Point: 128-131° Boiling Point: 186-187°/12mm Beilstein Reference: 107082 MDL No.: MFCD00134298

Synonyms:  2-Hydroxy-4-methylpyridine, 4-Methylpyridin-2-ol
  • CAS Number:13466-41-6
  • Formula:C₆H₇NO
  • MDL Number:MFCD00134298
  • Molecular Weight:109.13 g/mol
  • Boiling Pt:186…187 °C (12 mmHg)
  • Melting Pt:128…131 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 74,80 €
    74,80 €
    1 * 5 g
       74,80 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 249,00 €
    249,00 €
    1 * 25 g
       249,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.