
2-Hydroxy-5-methylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91914-06-6
Formula:C₆H₇NO
MDL Number:MFCD03092888
:109.13 g/mol
Melting Pt:181…183 °C
EINECS:213-713-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:180 to 185 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together