
Bis(4-chlorophenyl)sulphone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80-07-9
:
Formula:C₁₂H₈Cl₂O₂S
MDL Number:MFCD00000619
Boiling Pt:397 °C
Melting Pt:146…151 °C
Flash Pt:233 °C
:287.17 g/mol
EINECS:201-247-9
:GHS07
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:146 to 151 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together