
1-Bromo-2,3-dichlorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56961-77-4
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000536
Boiling Pt:243 °C
Melting Pt:56…60 °C
:225.9 g/mol
Storage Temperature:Ambient
EINECS:260-476-2
:GHS07
Related Information
Frequently Bought Together