Order Entry
Italy
Orders LinkContactUsLinkComponent
Counting chambers, Bürker
Supplier:   HIRSCHMANN
Counting chambers, Bürker
Supplier:  HIRSCHMANN
Certificates
Counting chambers, Bürker
Counting chambers, Bürker
4 Options Available Below
Product Details & Documents
These counting chambers with double net ruling are suitable for in vitro diagnostic applications.
 : In vitro Diagnostic Devices Directive (98/79/EC)
 : Includes two cover glasses.
Product Family Options
Product Information
  • Type
    Depth of chamber
    Availability
    Price
  • 612-5719
    Counting chamber
    Counting chamber
    Bürker
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 63,70 €
    63,70 €
    1 * 1 pezzi
       63,70 €
  • 612-5723
    Counting chamber, bright-lined
    Counting chamber, bright-lined
    Bürker
    0,10 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 151,00 €
    151,00 €
    1 * 1 pezzi
       151,00 €
  • 612-5727
    Counting chamber with clips
    Counting chamber with clips
    Bürker
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 88,10 €
    88,10 €
    1 * 1 pezzi
       88,10 €
  • 612-5729
    Counting chamber with clips, bright-lined
    Counting chamber with clips, bright-lined
    Bürker
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 169,00 €
    169,00 €
    1 * 1 pezzi
       169,00 €