Order Entry
Italy
ContactUsLinkComponent
Bromobenzene 99%, pure
Bromobenzene 99%, pure
Certificates
undefined
Bromobenzene 99%, pure

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5580 to 1.5600 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together