Order Entry
Italy
Orders LinkContactUsLinkComponent
1,2-Dibromobenzene 98%

1,2-Dibromobenzene 98%

Irritant
Environment
1,2-Dibromobenzene 98%
Certificates
1,2-Dibromobenzene 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:583-53-9
  • Formula:C₆H₄Br₂
  • MDL Number:MFCD00000057
  • Molecular Weight:235.91 g/mol
  • Boiling Pt:224 °C (760 mmHg)
  • Melting Pt:4…6 °C
  • Density:1.956 g/cm³
  • Flash Pt:91 °C
  • Storage Temperature:Ambient
  • EINECS:209-507-3
  • UN:3082
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow to light pink Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6100 to 1.6120 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 105,00 €
    105,00 €
    1 * 25 mL
       105,00 €

  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 298,00 €
    298,00 €
    1 * 100 mL
       298,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.