Anthracene for synthesis, Sigma-Aldrich®
: Merck
:120-12-7
:
:C₁₄H₁₀
:MFCD00001240
:178.23 g/mol
:13,00687
:340 °C (1013 hPa)
:111…112 °C
:1.24 g/cm³ (20 °C)
:Ambient
:204-371-1
:3077
:9,III
:M-Clarity™ quality level = MQ200
:121 °C (closed cup)
:GHS09
- Solubility:- 0.0001 g/l (20 °C)
- Melting Point:213 - 216 °C
- Molar Mass:178.23 g/mol
- Bulk Density:330 kg/m3
- Boiling Point:340 °C (1013 hPa)
- Vapor Pressure:0.000008 hPa (25 °C)
- Flash Point:121 °C
- Explosion Limit:0.6 %(V)
- Ignition Point:540 °C
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Frequently Bought Together