Order Entry
Italy
ContactUsLinkComponent
Anthracene
Anthracene
Catalog # EHERC20520000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Anthracene
Catalog # EHERC20520000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :178.23 g/mol
Merck Index:13,00687
Boiling Pt:340 °C (1013 hPa)
Melting Pt:111…112 °C
Density:1.24 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-371-1
 :GHS09
CAS Number:120-12-7
Formula:C₁₄H₁₀
MDL Number:MFCD00001240