Anthracene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120-12-7
:
Formula:C₁₄H₁₀
MDL Number:MFCD00001240
:178.23 g/mol
Merck Index:14,682
Boiling Pt:340…342 °C
Melting Pt:213…216 °C
Density:1.250
Flash Pt:121 °C (249 °F)
Storage Temperature:Ambient
EINECS:204-371-1
ADR:9,III
:GHS07
Related Information
Frequently Bought Together