Anthracene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120-12-7
:
Formula:C₁₄H₁₀
MDL Number:MFCD00001240
:178.23 g/mol
Merck Index:13,00687
Boiling Pt:340 °C (760 mmHg)
Melting Pt:215…218 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:121 °C
Storage Temperature:Ambient
EINECS:204-371-1
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to yellow to beige to green-beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:214 to 219 °C
- GC:≥98.5 %
- Impurity:≤0.05 % Alkali and other salts (as sulfates)
- Acidity (CH3COOH):≤0.001 %
Specification Test Results
Frequently Bought Together