2-Methyl-2-butene
Catalog # 8.20780.9040
Supplier: Merck
CAS Number:
Specifications
- Pk:40 kg
- Seq:4367
Specifications
About this item
:
:70.13 g/mol
Merck Index:12,00649
Boiling Pt:39 °C (1013 hPa)
Melting Pt:–124 °C
Density:0.6623 g/cm³ (20 °C)
Flash Pt:–45 °C
EINECS:208-156-3
UN:2460
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Refrigerator
CAS Number:513-35-9
:26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276