Jump to:
About this item
- CAS Number:592-41-6
- Formula:C₆H₁₂
- MDL Number:MFCD00009505
- Molecular Weight:84.16 g/mol
- Boiling Pt:63 °C (1013 hPa)
- Melting Pt:–140 °C
- Density:0.675 g/cm³ (20 °C)
- Flash Pt:–26 °C
- Storage Temperature:Ambient
- EINECS:209-753-1
- UN:2370
- ADR:3,II
5 Options Available Below
Product Details & Documents
- CAS Number:592-41-6
- Formula:C₆H₁₂
- MDL Number:MFCD00009505
- Molecular Weight:84.16 g/mol
- Boiling Pt:63 °C (1013 hPa)
- Melting Pt:–140 °C
- Density:0.675 g/cm³ (20 °C)
- Flash Pt:–26 °C
- Storage Temperature:Ambient
- EINECS:209-753-1
- UN:2370
- ADR:3,II
Documents
msds_820639
SDS
ProductPage_820639 ProductPage_820639
Product specification
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



