Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hexene ≥98%

1-Hexene ≥98%

Flammable
Health
   Sustainable Options Available
Nome del fornitore:   Thermo Fisher Scientific
1-Hexene ≥98%
Nome del fornitore:  Thermo Fisher Scientific
   Sustainable Options Available
Certificates
1-Hexene ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 592-41-6 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -25°(-13°F) Refractive Index: 1.3880 Odour: Mild Melting Point: -140° Boiling Point: 62-64° Beilstein Reference: 1209240 UN No.: 2370 MDL No.: MFCD00009505

  • CAS Number:592-41-6
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009505
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:62…64 °C
  • Melting Pt:–140 °C
  • Density:0.673
  • Flash Pt:–25 °C (–13 °F)
  • Storage Temperature:Ambient
  • EINECS:209-753-1
  • UN:2370
  • ADR:3,II
Specifications for Product Family

Specifications

Product Family Options

Product Information