Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1-Hexene 97%

1-Hexene 97%
 
1-Hexene 97%
1-Hexene 97%

 

 
  •  :592-41-6
  •  :C₆H₁₂
  •  :MFCD00009505
  •  :84.16 g/mol
  •  :62…65 °C (760 mmHg)
  •  :–140 °C
  •  :0.678 g/cm³
  •  :–26 °C
  •  :Ambient
  •  :209-753-1
  •  :2370
  •  :3,II
 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3867 to 1.3887 (20°C, 589 nm)
  • GC:
    ≥96.0 %
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - 27,00 €
    27,00 €
    1 * 100 mL
       27,00 €

  • Glass bottle
    1 L
       
       
    1 * 1 L
    1 * 1 L - 50,20 €
    50,20 €
    1 * 1 L
       50,20 €

  • Glass bottle
    2,5 L
       
       
    1 * 2,5 L
    1 * 2,5 L - 98,00 €
    98,00 €
    1 * 2,5 L
       98,00 €

  • Metal drum
    25 L
       
       
    1 * 25 L
    1 * 25 L - 597,00 €
    597,00 €
    1 * 25 L
       597,00 €