
cis-4-Heptenal 96%
Catalog # ACRO298220100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4320 to 1.4360 (20°C, 589 nm)
- GC:≥95 %
- Specific gravity:(25°C) 0.843 to 0.855
Specification Test Results
About this item
:
:112.17 g/mol
Boiling Pt:46 °C (13 mmHg)
Density:0.849 g/cm³
Flash Pt:37 °C
Storage Temperature:Ambient
EINECS:229-779-7
UN:1989
ADR:3,III
:GHS02
CAS Number:6728-31-0
Formula:C₇H₁₂O
MDL Number:MFCD00014682