2-Bromopropene 99% stabilised
: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide
:557-93-7
:
:C₃H₅Br
:MFCD00000140
:120.98 g/mol
:47…49 °C (760 mmHg)
:–125 °C
:1.36 g/cm³
:4 °C
:Refrigerator
:209-185-4
:1993
:3,II
:GHS07
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4420 to 1.4460 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:Approx. 100 ppm BHT
Frequently Bought Together