Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dimethylhexane, Supelco®

3,4-Dimethylhexane, Supelco®

Glass bottle, 1 mL
Codice catalogo 40512-1ML-F
Flammable
Health
Irritant
Environment
Nome del fornitore:   Merck
3,4-Dimethylhexane, Supelco®
Codice catalogo 40512-1ML-F
Nome del fornitore:  Merck
Numero CAS:  583-48-2
3,4-Dimethylhexane, Supelco®
3,4-Dimethylhexane, Supelco®

Glass bottle, 1 mL

Numero CAS:  583-48-2
Price
44,90 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • CAS Number:583-48-2
  • Formula:C₈H₁₈
  • MDL Number:MFCD00039927
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:118 °C (1013 hPa)
  • Melting Pt:–64 °C
  • Density:0.7152 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-504-7
  • UN:1262
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.