



2,4-Dimethylpentane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-08-7
:
Formula:C₇H₁₆
MDL Number:MFCD00008945
:100.2 g/mol
Boiling Pt:79…80 °C
Melting Pt:∼–123 °C
Density:0.670
Flash Pt:–12 °C (10 °F)
EINECS:203-548-0
UN:1206
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together