
2-Chloro-2-methylpropane for synthesis, Sigma-Aldrich®
: Merck
:507-20-0
:
:C₄H₉Cl
:MFCD00000816
:92.57 g/mol
:13,01561
:51 °C (1013 hPa)
:–28 °C
:0.85 g/cm³ (20 °C)
:Refrigerator
:208-066-4
:1127
:3,II
:M-Clarity™ quality level = MQ200
:–27 °C
:GHS02
- Solubility:(20 °C) insoluble,(slow decomposition)
- Melting Point:-28 °C
- Molar Mass:92.57 g/mol
- Boiling Point:51 °C (1013 hPa)
- Vapor Pressure:350 hPa (20 °C)
- Flash Point:-27 °C
- Refractive Index:1.3848 (20 °C, 589 nm)
- Explosion Limit:1.8 - 10.1 %(V)
- Density:0.84 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:540 °C
Frequently Bought Together