

4-Methyl-2-pentanone ≥98.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-10-1
:
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
:100.16 g/mol
Merck Index:14,5207
Boiling Pt:117…118 °C
Melting Pt:–84 °C
Density:0.801
Flash Pt:13 °C (55 °F)
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Related Information
Frequently Bought Together