2,6-Dimethyl-4-heptanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Diisobutyl Ketone
CAS Number:108-83-8
:
Formula:C₉H₁₈O
MDL Number:MFCD00008940
:142.24 g/mol
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–46 °C
Density:0.806 g/cm³ (20 °C)
Flash Pt:60 °C
UN:1157
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:203-620-1
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.5 g/l (20 °C)
- Melting Point:-41 °C
- Molar Mass:142.24 g/mol
- Boiling Point:163 - 173 °C (1013 hPa)
- Vapor Pressure:1.6 hPa (20 °C)
- Flash Point:47 °C
- Refractive Index:1.4143 (20 °C, 589 nm)
- Explosion Limit:0.8 - 6.2 %(V)
- Density:0.81 g/cm3 (20 °C)
- Ignition Point:345 °C
- Saturation concentration:9 g/m3 (20 °C) Air
- Evaporation number:48
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Frequently Bought Together