

2,6-Dimethyl-4-heptanone
Catalog # EHERC12727300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:142.24 g/mol
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–46 °C
Density:0.806 g/cm³ (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:203-620-1
UN:1224
ADR:3,III
:GHS02
:GHS07
CAS Number:108-83-8
Formula:C₉H₁₈O
MDL Number:MFCD00008940