Order Entry
Italy
ContactUsLinkComponent
2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone
Irritant
Flammable
2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone
Certificates
undefined
Irritant
Flammable
2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4100 to 1.4180 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥94 % (main compound + 4.6-Dimethyl-2-heptanone)

Specification Test Results

Related Information

Frequently Bought Together