

2,4-Dimethyl-3-pentanone for synthesis
Catalog # 8.22037.9016
Supplier: Merck
CAS Number:
Specifications
- Pk:16 kg
- Seq:4226
Specifications
About this item
:
:114.19 g/mol
Boiling Pt:124 °C (1013 hPa)
Melting Pt:–69 °C
Density:0.806 g/cm³ (20 °C)
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:209-294-7
UN:1224
ADR:3,II
:GHS02
:GHS07
CAS Number:565-80-0
Formula:C₇H₁₄O
MDL Number:MFCD00008918