Order Entry
Italy
ContactUsLinkComponent
1-Bromo-2-methylpropane for synthesis, Sigma-Aldrich®
Flammable
1-Bromo-2-methylpropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
1-Bromo-2-methylpropane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    -118 °C
  • Molar Mass:
    137.01 g/mol
  • Boiling Point:
    92 °C (1013 hPa)
  • Vapor Pressure:
    41 hPa (20 °C)
  • Flash Point:
    19 °C
  • Refractive Index:
    1.435 (20 °C, 589 nm)
  • Density:
    1.26 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together