
n-Pentadecane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-62-9
:
Formula:H₃C(CH₂)₁₃CH₃
MDL Number:MFCD00008990
:212.42 g/mol
Boiling Pt:271 °C (1013 hPa)
Melting Pt:13…15 °C
Density:0.768 g/cm³ (20 °C)
Flash Pt:132 °C
UN:3295
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:211-098-1
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:10 °C
- Molar Mass:212.41 g/mol
- Boiling Point:270 °C (1013 hPa)
- Vapor Pressure:- 0.2 hPa (20 °C)
- Flash Point:132 °C
- Explosion Limit:0.45 - 6.5 %(V)
- Density:0.77 g/cm3 (20 °C)
- Viscosity Kinematic:- 7 mm2/s (40 °C)
Specification Test Results
Frequently Bought Together