Order Entry
Italy
ContactUsLinkComponent
Propargyl chloride 70% ∼9.2 mol/L in toluene
Flammable
Corrosive
Toxic
Health
Propargyl chloride 70% ∼9.2 mol/L in toluene
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Toxic
Health
Propargyl chloride 70% ∼9.2 mol/L in toluene
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Concentration (HNMR):
    70.0 to 77.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together