
ω,4-Dibromoacetophenone, Millipore®
Catalog # 361548-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:277.94 g/mol
Merck Index:13,01412
Boiling Pt:310 °C (1013 hPa)
Melting Pt:107…108 °C
Storage Temperature:Refrigerator
EINECS:202-783-6
UN:3261
ADR:8,II
:GHS05
CAS Number:99-73-0
Formula:C₈H₆Br₂O
MDL Number:MFCD00000200