

1-Methoxy-2,2,2-trifluoroethanol tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:431-46-9
:
Formula:C₃H₅F₃O₂
MDL Number:MFCD00013572
Boiling Pt:96 °C
Density:1.360
Flash Pt:42 °C (107 °F)
EINECS:207-072-4
UN:3271
ADR:3,III
:130.07 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together