4,4,4-Trifluoro-1-phenyl-1,3-butanedione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:326-06-7
:
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD00000425
:216.16
Boiling Pt:224 °C
Melting Pt:37…40 °C
Density:1.113
Flash Pt:98 °C (208 °F)
Storage Temperature:Ambient
EINECS:206-307-8
Frequently Bought Together