D-α-Tocopherol 97+%
Glass bottle for solids, 100 mg
: (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
- :59-02-9
- :C₂₉H₅₀O₂
- :MFCD00072045
- :430.71 g/mol
- :200…220 °C (.1 mmHg)
- :2.5…3.5 °C
- :>110 °C
- :Refrigerator
- :233-466-0
- Appearance:Clear yellow to red-brown Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.5050 to 1.5070 (20°C, 589 nm)
- GC:≥97.0 %
- Pack type:Glass bottle for solids
- Pk:100 mg



