
trans,trans-1,4-Diphenyl-1,3-butadiene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:538-81-8
:886-65-7
Formula:C₁₆H₁₄
MDL Number:MFCD00004791
:206.29 g/mol
Melting Pt:151…154 °C
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:151 to 154 °C
- GC:≥98.5 %
- Freezing point:2.0 to 3.0 °C
Specification Test Results
Frequently Bought Together