n-Nonane
Catalog # EHERC15623000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–53 °C
Density:0.718 g/cm³ (20 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:203-913-4
UN:1920
ADR:3,III
:128.26 g/mol
:GHS02
:GHS08
CAS Number:111-84-2
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574