Order Entry
Italy
ContactUsLinkComponent
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Toxic
Environment
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Toxic
Environment
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    Min. 97 %
  • Acidity:
    Max. 0.0003 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 50 ppm
  • Absorbance (210 nm):
    Max. 0.3
  • Absorbance (220 nm):
    Max. 0.1
  • Absorbance (245 nm):
    Max. 0.01
  • Transmittance (210 nm):
    Min. 50 %
  • Transmittance (220 nm):
    Min. 80 %
  • Transmittance (245 nm):
    Min. 98 %

Specification Test Results

Frequently Bought Together