Order Entry
Italy
ContactUsLinkComponent
 
1,3-Propanedithiol 98%
 
undefined
1,3-Propanedithiol 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5380 to 1.5420 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Density:
    (20°C) 1.038 to 1.048 g/ml

 

 

Frequently Bought Together