Order Entry
Italy
Orders LinkContactUsLinkComponent
1,7-Heptanediol 95%

1,7-Heptanediol 95%

1,7-Heptanediol 95%
Certificates
1,7-Heptanediol 95%

2 Options Available Below

Product Details & Documents
Synonyms:  1,7-Dihydroxy heptane
  • CAS Number:629-30-1
  • Formula:C₇H₁₆O₂
  • MDL Number:MFCD00002987
  • Molecular Weight:132.2 g/mol
  • Boiling Pt:259 °C (760 mmHg)
  • Melting Pt:17…19 °C
  • Density:0.951 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:211-085-0

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4535 to 1.4565 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Specific optical rotation:
    -5.5° to -7.9° (20°C, 589 nm) (c=1,CHCl3)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 33,00 €
    33,00 €
    1 * 1 g
       33,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 97,40 €
    97,40 €
    1 * 10 g
       97,40 €