Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific
About this item
Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odour: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885
Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
- CAS Number:107-21-1
- Formula:HOCH₂CH₂OH
- MDL Number:MFCD00002885
- Molecular Weight:62.07 g/mol
- Merck Index:14,3798
- Boiling Pt:196…198 °C
- Melting Pt:–13 °C
- Density:1.113
- Flash Pt:119 °C (246 °F)
- Storage Temperature:Ambient
- EINECS:203-473-3
- REACH:01-2119456816-28
4 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odour: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885
Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
- CAS Number:107-21-1
- Formula:HOCH₂CH₂OH
- MDL Number:MFCD00002885
- Molecular Weight:62.07 g/mol
- Merck Index:14,3798
- Boiling Pt:196…198 °C
- Melting Pt:–13 °C
- Density:1.113
- Flash Pt:119 °C (246 °F)
- Storage Temperature:Ambient
- EINECS:203-473-3
- REACH:01-2119456816-28
Documents
Certificates
Search by Lot NumberALFAA11591.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- A11591.30
- A11591.36
- A11591.0E
- A11591.0C
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.A11591.30Environmentally PreferableEnvironmental certification - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.A11591.36Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.A11591.0EEnvironmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.A11591.0CEnvironmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health



