


Methanol, anhydrous (max. 0.005% H₂O) ≥99.8% for synthesis
Catalog # 20856.296
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Appearance:Clear colourless liquid
- Colour value:Max. 10 APHA
- Assay (calculated on anhydrous):Min. 99.8 %
- Acidity (as HCOOH):Max. 0.0010 %
- Alkalinity (as NH3):Max. 0.00010 %
- Acetone + aldehydes (as CH3COCH3):Max. 0.0010 %
- Formaldehyde:Max. 0.0001 %
- Residue on evaporation:0.0010 %
- Reducing substances:Max. 0.0002 %
- Water (K.F.):Max. 0.0050 %
- Substances discoloured by H2SO4:Max. 20 APHA
- Ethanol:Max. 0.100 %
- Cl (Chloride):Max. 0.5 ppm
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.5 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.01 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
About this item
:
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595