


Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Appearance:Clear colourless liquid
- Assay (on anhydrous substance):Min. 99.8 %
- IR Spectrum:Passes test
- Solubility in water:Passes test ACS
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.00006 meq/g
- Boiling point:64 - 65 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.791 - 0.792
- Density (20/20):0.791 - 0.793
- Substances coloured by H2SO4:Max. 10 APHA
- Acetone + aldehydes (as CH3COCH3):Max. 10 ppm
- Acetone:Max. 5 ppm
- Acetaldehyde:Max. 5 ppm
- Ethanol:Max. 0.1 %
- Evaporation residue:Max. 10 ppm
- Formaldehyde:Max. 1 ppm
- Substances reducing KMnO4 (as O):Max. 2 ppm
- Water:Max. 0.05 %
- Cl (Chloride):Max. 0.5 ppm
- Al (Aluminium):Max. 0.1 ppm
- B (Boron):Max. 0.08 ppm
- Ba (Barium):Max. 0.05 ppm
- Ca (Calcium):Max. 0.8 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.01 ppm
- Na (Sodium):Max. 0.7 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.2 ppm
- Absorbance (225 nm):Max. 0.17
- Transmittance (210 nm):Min. 20 %
- Transmittance (220 nm):Min. 50 %
- Transmittance (225 nm):Min. 68 %
- Transmittance (250 nm):Min. 95 %
- Conforms to ACS:Passes test
- Conforms Ph.Eur. R 1053200:Passes test
- Conforms Ph.Eur. R1 1053201:Passes test
- Conforms Ph.Eur. R2 1053202:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250)
Frequently Bought Together