Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Heneicosanol
1-Heneicosanol
1-Heneicosanol
EHERC14085200
:
EHRENSTORFER
:
1-Heneicosanol
EHERC14085200
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
312.58 g/mol
:
170 °C (1013 hPa)
:
68…69 °C
:
Refrigerator
:
239-673-2
:
15594-90-8
:
H₃C(CH₂)₁₉CH₂OH
:
MFCD00062834