1-Docosanol for synthesis, Sigma-Aldrich®
Catalog # 8.22047.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:68 - 71 °C
- Molar Mass:326.6 g/mol
- Boiling Point:180 °C (0.3 hPa)
- Synonyms:Behenyl alcohol
- Hill Formula:C22H46O
Specification Test Results
About this item
:
:326.61 g/mol
Merck Index:13,03433
Boiling Pt:376 °C (1013 hPa)
Melting Pt:68…71 °C
Density:∼0.81 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-546-6
CAS Number:661-19-8
Formula:H₃C(CH₂)₂₀CH₂OH
MDL Number:MFCD00002939