Tridodecylamine (hydrogen ionophore I) for synthesis, Sigma-Aldrich®
8.21160.9020
: Merck
:
- Pk:20 kg
- Seq:4241
- Solubility:(20 °C) insoluble
- Melting Point:16 °C
- Molar Mass:522.01 g/mol
- Boiling Point:450 °C
- Flash Point:190 °C
- Density:0.82 g/cm3 (20 °C)
- Ignition Point:245 °C
:
:522 g/mol
:450 °C (1013 hPa)
:16 °C
:0.82 g/cm³ (20 °C)
:190 °C
:Ambient
:203-063-4
:3082
:9,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
:102-87-4
:C₃₆H₇₅N
:MFCD00008971