Iodoethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-03-6
:
Formula:C₂H₅I
MDL Number:MFCD00001091
:155.97 g/mol
Merck Index:13,03846
Boiling Pt:72 °C (1013 hPa)
Melting Pt:–108 °C
Density:1.936 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:53 °C
:Stabilised with silver
Storage Temperature:Ambient
EINECS:200-833-1
UN:1993
:GHS02
:GHS07
Specification Test Results
- Solubility:4 g/l (20 °C) (slow decomposition)
- Melting Point:-108 °C
- Molar Mass:155.97 g/mol
- Boiling Point:69 - 73 °C (1013 hPa)
- Vapor Pressure:133 hPa (18 °C)
- Flash Point:53 °C
- Refractive Index:1.513 (20 °C, 589 nm)
- Density:1.93 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together