

Diiodomethane 99+% stabilised
:75-11-6
:
:CH₂I₂
:MFCD00001079
:267.84 g/mol
:12,06143
:181 °C (760 mmHg)
:6 °C
:3.32 g/cm³
:>110 °C
:2810
:6.1,III
:stabilised with silver wire
:200-841-5
:GHS05
:GHS06
:Ambient
- Appearance (Color):Clear colorless to yellow to red to orange-brown
- Appearance (Form):Liquid
- Infrared spectrum:Conforms
- GC:≥99.0 %
- Stabilizer:Silver
Frequently Bought Together