Order Entry
Italy
ContactUsLinkComponent
1-Iodobutane for synthesis, Sigma-Aldrich®
Flammable
Toxic
1-Iodobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
1-Iodobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.2 g/l (20 °C)
  • Melting Point:
    -103 °C
  • Molar Mass:
    184.02 g/mol
  • Boiling Point:
    130 °C (1013 hPa)
  • Vapor Pressure:
    18.5 hPa (25 °C)
  • Flash Point:
    33 °C
  • Refractive Index:
    1.4979 (20 °C, 589 nm)
  • Density:
    1.62 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together