Order Entry
Italy
ContactUsLinkComponent
1-Chloropropane
Flammable
Irritant
1-Chloropropane
Catalog # EHERCA11502500
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Irritant
1-Chloropropane
Catalog # EHERCA11502500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :78.54 g/mol
Merck Index:12,08032
Boiling Pt:46 °C (1013 hPa)
Melting Pt:–122.8 °C
Density:1.3914 g/cm³ (20 °C)
Flash Pt:–31 °C
Storage Temperature:Refrigerator
EINECS:208-749-7
UN:1278
ADR:3,II
 :GHS02
 :GHS07
CAS Number:540-54-5
Formula:C₃H₇Cl
MDL Number:MFCD00000995