Order Entry
Italy
ContactUsLinkComponent
1,4-Dichlorobutane for synthesis, Sigma-Aldrich®
Flammable
1,4-Dichlorobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
1,4-Dichlorobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2.4 g/l (20 °C)
  • Melting Point:
    -38 °C
  • Molar Mass:
    127.01 g/mol
  • Boiling Point:
    155 °C (1013 hPa)
  • Vapor Pressure:
    5 hPa (20 °C)
  • Flash Point:
    53 °C
  • Refractive Index:
    1.4545 (20 °C, 589 nm)
  • Explosion Limit:
    1.8 - 8.9 %(V)
  • Density:
    1.14 g/cm3 (20 °C)
  • pH:
    7.3 (H2O, 20 °C)
  • Ignition Point:
    220 °C

Specification Test Results

Related Information

Frequently Bought Together