1,2-Dichloropropane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-87-5
:
Formula:ClH₂CCH₂CH₂Cl
MDL Number:MFCD00000868
:112.99 g/mol
Merck Index:14,7854
Boiling Pt:95…97 °C
Melting Pt:–100 °C
Density:1.157
Flash Pt:4 °C (39 °F)
Storage Temperature:Ambient
EINECS:201-152-2
UN:1279
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together