

Citric acid, anhydrous ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
:192.13 g/mol
Merck Index:14,2326
Boiling Pt:310 °C (1013 hPa)
Melting Pt:153…154 °C
Density:1.665
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS05
:GHS07
Related Information
Frequently Bought Together